Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

462063

Sigma-Aldrich

(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde

95%

Synonym(s):

(+)-N-Boc-N,O-isopropylidene-D-serinal, tert-Butyl (R)-(+)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO4
CAS Number:
Molecular Weight:
229.27
Beilstein:
3651554
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

optical activity

[α]23/D +90°, c = 1 in chloroform

impurities

<6% methyl (R)-(+)-3-(tert-butoxycarbonyl)-2,2-dimethyl-4-oxazolidinecarboxylate (CAS 95715-87-0)

refractive index

n20/D 1.445 (lit.)

bp

67 °C/0.3 mmHg (lit.)

density

1.06 g/mL at 25 °C (lit.)

functional group

aldehyde
ether

SMILES string

[H]C(=O)[C@H]1COC(C)(C)N1C(=O)OC(C)(C)C

InChI

1S/C11H19NO4/c1-10(2,3)16-9(14)12-8(6-13)7-15-11(12,4)5/h6,8H,7H2,1-5H3/t8-/m0/s1

InChI key

PNJXYVJNOCLJLJ-QMMMGPOBSA-N

Application

Employed in the synthesis of fluorinated analogs of glutemic and glutamine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Dave et al.
Amino acids, 24(3), 245-261 (2003-04-23)
Gamma-fluorinated analogues of glutamic acid and glutamine are compounds of biological interest. Syntheses of such compounds are extensively reviewed in this article. 4-fluoroglutamic acid was prepared as a mixture of racemic diastereomers by Michael reaction, inverse-Michael reaction or by electrophilic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service