Skip to Content
MilliporeSigma
All Photos(1)

Documents

460427

Sigma-Aldrich

Neodymium(III) acetylacetonate hydrate

Synonym(s):

2,4-Pentanedione neodymium(III) derivative, Nd(acac)3, Neodymium(III) 2,4-pentanedionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nd(C5H7O2)3 · xH2O
CAS Number:
Molecular Weight:
441.56 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

composition

Nd, 30.0-34.0% EDTA titration

reaction suitability

core: neodymium
reagent type: catalyst

mp

143 °C (dec.) (lit.)

SMILES string

O.CC(=O)\C=C(\C)O[Nd](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Nd.H2O/c3*1-4(6)3-5(2)7;;/h3*3,6H,1-2H3;;1H2/q;;;+3;/p-3/b3*4-3-;;

InChI key

ZOWCSWGOVNMTLL-KJVLTGTBSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ruiyang Kang et al.
Scientific reports, 9(1), 9135-9135 (2019-06-27)
With the development of electronic devices such as integrated circuits toward the continual increase in power density and consumption, the efficient heat dissipation and low thermal expansion of materials become one of the most important issue. However, conventional polymers have

Articles

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service