Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

460125

Sigma-Aldrich

Poly(ethylene glycol) dioleate

average Mn ~914

Synonym(s):

O,O′-Dioleoylpolyethylene glycol 400, Polyethylene glycol 400 dioleate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7CO(OCH2CH2)nO2C(CH2)7CH=CH(CH2)7CH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~914

refractive index

n20/D 1.471 (lit.)

bp

>260 °C (lit.)

mp

−15 °C (lit.)

acid number

4 mg KOH/g

hydroxyl value

10 mg KOH/g

iodine value

75

solubility

toluene, ethanol and acetone: soluble (dispersible in water)

density

0.945 g/mL at 25 °C (lit.)

SMILES string

OCCO.[H]\C(CCCCCCCC)=C(/[H])CCCCCCCC(O)=O

InChI

1S/C38H70O4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-37(39)41-35-36-42-38(40)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20H,3-16,21-36H2,1-2H3/b19-17-,20-18-

InChI key

NKSOSPOXQKNIKJ-CLFAGFIQSA-N

Application

Plasticizer

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service