457876
Poly(ethylene glycol) methyl ether methacrylate solution
average Mn 2,000, 50 wt. % in H₂O, methacrylate, methoxy
Synonym(s):
Polyethylene glycol
About This Item
Recommended Products
Product Name
Poly(ethylene glycol) methyl ether methacrylate solution, average Mn 2,000, 50 wt. % in H2O
mol wt
average Mn 2,000
Quality Level
reaction suitability
reagent type: chemical modification reagent
reaction type: Polymerization Reactions
concentration
50 wt. % in H2O
refractive index
n20/D 1.4054
density
1.079 g/mL at 25 °C
Ω-end
methacrylate
α-end
methoxy
polymer architecture
shape: linear
functionality: monofunctional
SMILES string
COCCOCCOCCOCCOC(=O)C(C)=C
InChI
1S/C13H24O6/c1-12(2)13(14)19-11-10-18-9-8-17-7-6-16-5-4-15-3/h1,4-11H2,2-3H3
InChI key
KRCGBOKYIUDIFY-UHFFFAOYSA-N
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.
Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.
Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.
Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service