Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

457396

Sigma-Aldrich

4-Fluoroindole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6FN
CAS Number:
Molecular Weight:
135.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

bp

90 °C/0.4 mmHg (lit.)

mp

30-32 °C (lit.)

functional group

fluoro

SMILES string

Fc1cccc2[nH]ccc12

InChI

1S/C8H6FN/c9-7-2-1-3-8-6(7)4-5-10-8/h1-5,10H

InChI key

ZWKIJOPJWWZLDI-UHFFFAOYSA-N

Application

  • Reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Reactant for preparation of antifungal agents
  • Reactant for preparation of Sodium-Dependent Glucose Co-transporter 2 (SGLT2) Inhibitors for the Management of Hyperglycemia in Diabetes
  • Reactant for preparation of Potent Selective Serotonin Reuptake Inhibitors
  • Reactant for preparation of Inhibitors of HIV-1 attachment
  • Reactant for preparation of monoamine reuptake inhibitors
  • Reactant for preparation of histone deacetylase (HDAC) inhibitors
  • Reactant for preparation of inhibitors of proliferation of human breast cancer cells

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Synthesis of (L)-4-Fluorotryptophan.
Konas DW, et al.
Synthetic Communications, 42(1), 144-152 (2012)
Journal of the Chemical Society. Perkin Transactions 1, 1765-1765 (1994)
(Erratum)
Journal of Medicinal Chemistry, 2442-2442 null
A synthesis of (-)-indolactam V.
Semmelhack MF and Rhee H.
Tetrahedron Letters, 34(39), 1395-1398 (1993)
4-Fluoroindole and Derivatives.
Bentov M, et al.
Israel J. Chem., 2(1), 25-28 (1964)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service