Skip to Content
MilliporeSigma
All Photos(1)

Documents

456144

Sigma-Aldrich

(S)-(−)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11ClO2
CAS Number:
Molecular Weight:
150.60
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

description

Specific optical acitvity ([α]24/D ·42.1°, neat)

Quality Level

Assay

98%

refractive index

n20/D 1.434 (lit.)

bp

63 °C/37 mmHg (lit.)

density

1.103 g/mL at 25 °C (lit.)

SMILES string

CC1(C)OC[C@@H](CCl)O1

InChI

1S/C6H11ClO2/c1-6(2)8-4-5(3-7)9-6/h5H,3-4H2,1-2H3/t5-/m1/s1

InChI key

BNPOTXLWPZOESZ-RXMQYKEDSA-N

Application

(S)-(−)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane can be used as a building block in the synthesis of hydroxylated polyamine analogs of biological importance.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and evaluation of hydroxylated polyamine analogues as antiproliferatives
Bergeron RJ, et al.
Journal of Medicinal Chemistry, 43(2), 224-235 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service