Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

454370

Sigma-Aldrich

6-Amino-2-mercaptobenzothiazole

97%

Synonym(s):

6-Amino-2-benzothiazolethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2S2
CAS Number:
Molecular Weight:
182.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

267-272 °C (lit.)

SMILES string

Nc1ccc2nc(S)sc2c1

InChI

1S/C7H6N2S2/c8-4-1-2-5-6(3-4)11-7(10)9-5/h1-3H,8H2,(H,9,10)

InChI key

IDPNFKLUBIKHSW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antimicrobial activity of 2-alkylthio-6-aminobenzothiazoles.
Sidoova E, et al.
Chemical Papers, 33, 830-836 (1979)
[A QSAR study of synthetic stimulators of growth of domestic animals on the basis of 6-amino-2-mercaptobenzothiazole].
P Zahradník et al.
Ceskoslovenska farmacie, 37(10), 440-442 (1988-12-01)
Electrochemical Characterization of Gold 6-Amino-2-mercaptobenzothiazole Self-Assembled Monolayer for Dopamine Detection in Pharmaceutical Samples.
Shervedani RK, et al.
Electroanalysis, 22(9), 969-977 (2010)
Triorganotin (IV) complexes of Schiff base derived from 6-amino-2-mercaptobenzothiazole: Synthesis, characterization and X-ray crystal structures.
Ma C, et al.
Inorgorganica Chimica Acta, 360(7), 2439-2446 (2007)
Aiming Sun et al.
ChemMedChem, 6(4), 654-666 (2011-03-03)
Small molecules, namely coactivator binding inhibitors (CBIs), that block estrogen signaling by directly inhibiting the interaction of the estrogen receptor (ER) with coactivator proteins act in a fundamentally different way to traditional antagonists, which displace the endogenous ligand estradiol. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service