Skip to Content
MilliporeSigma
All Photos(1)

Documents

451274

Sigma-Aldrich

Scandium(III) chloride hexahydrate

99.999% trace metals basis

Synonym(s):

Scandium trichloride hexahydrate, Scandium(3+) trichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ScCl3 · 6H2O
CAS Number:
Molecular Weight:
259.41
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

powder

impurities

≤15.0 ppm Trace Rare Earth Analysis

mp

63-66 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Sc](Cl)Cl

InChI

1S/3ClH.6H2O.Sc/h3*1H;6*1H2;/q;;;;;;;;;+3/p-3

InChI key

FKZFOHABAHJDIK-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mizuho Koike et al.
Nature communications, 11(1), 1988-1988 (2020-04-26)
Understanding the origin of organic material on Mars is a major issue in modern planetary science. Recent robotic exploration of Martian sedimentary rocks and laboratory analyses of Martian meteorites have both reported plausible indigenous organic components. However, little is known
Yingying Li et al.
Frontiers in chemistry, 8, 832-832 (2020-11-12)
The rare-earth nanocrystals containing Er3+ emitters offer very promising tools for imaging applications, as they can not only exhibit up-conversion luminescence but also down-conversion luminescence in the second near-infrared window (NIR II). Doping non-lanthanide cations into host matrix was demonstrated

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service