Skip to Content
MilliporeSigma
All Photos(1)

Documents

448621

Sigma-Aldrich

1,1,3,3,5,5-Hexamethyltrisiloxane

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2SiHO]2Si(CH3)2
CAS Number:
Molecular Weight:
208.48
Beilstein:
1746341
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.383 (lit.)

bp

128 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

SMILES string

[H][Si](C)(C)O[Si](C)(C)O[Si]([H])(C)C

InChI

1S/C6H20O2Si3/c1-9(2)7-11(5,6)8-10(3)4/h9-10H,1-6H3

InChI key

HZBDPZBVINJJET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liu Yang et al.
Sensors (Basel, Switzerland), 15(6), 12884-12890 (2015-06-05)
Hydrogen-bond acidic polymers are important high affinity materials sensitive to organophosphates in the chemical warfare agent sensor detection process. Interactions between the sensor sensitive materials and chemical warfare agent simulants were studied by inverse gas chromatography. Hydrogen bonded acidic polymers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service