Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

445908

Sigma-Aldrich

Poly(ethylene glycol) dimethyl ether

average MN 2,000, cross-linking reagent click chemistry

Synonym(s):

Polyethylene glycol, Dimethyl polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)nCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

Poly(ethylene glycol) dimethyl ether, average Mn ~2,000

form

solid

Quality Level

mol wt

average Mn ~2,000

reaction suitability

reagent type: cross-linking reagent
reaction type: click chemistry

bp

>250 °C

transition temp

Tm 53 °C

density

1.08 g/mL at 25 °C

polymer architecture

shape: linear
functionality: homobifunctional

SMILES string

CO.OCCO

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

InChI key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Paper Title: Molecular dynamics simulations for self-diffusion coefficients: The case of CO2, n-alkanes, and poly(ethylene glycol) dimethyl ethers - Studies self-diffusion coefficients using molecular dynamics simulations to provide insights into the behavior of poly(ethylene glycol) dimethyl ethers (OA Moultos et al., 2016).


Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

312.8 °F - closed cup

Flash Point(C)

156.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shravanthi Rajasekar et al.
Advanced materials (Deerfield Beach, Fla.), 32(46), e2002974-e2002974 (2020-10-02)
Despite the complexity and structural sophistication that 3D organoid models provide, their lack of vascularization and perfusion limit the capability of these models to recapitulate organ physiology effectively. A microfluidic platform named IFlowPlate is engineered, which can be used to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service