Skip to Content
MilliporeSigma

445347

(R)-(−)-1-Aminoindane

97%

Synonym(s):

(R)-(−)-1-Indanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H11N
CAS Number:
Molecular Weight:
133.19
UNSPSC Code:
12352116
NACRES:
NA.22
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3196204
Assay:
97%
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Drug Control: Új pszichoaktív anyag / New psychoactive substance (Hungary), 78/2022. (XII. 28.) BM rendelet.

Quality Segment

assay

97%

form

liquid

optical activity

[α]20/D −16.5°, c = 1.5 in methanol

drug control

Új pszichoaktív anyag / New psychoactive substance (Hungary), 78/2022. (XII. 28.) BM rendelet

refractive index

n20/D 1.562 (lit.)

bp

96-97 °C/8 mmHg (lit.)

mp

15 °C (lit.)

density

1.038 g/mL at 25 °C (lit.)

SMILES string

N[C@@H]1CCc2ccccc12

InChI

1S/C9H11N/c10-9-6-5-7-3-1-2-4-8(7)9/h1-4,9H,5-6,10H2/t9-/m1/s1

InChI key

XJEVHMGJSYVQBQ-SECBINFHSA-N

Gene Information

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Signal Word

Warning

Pictograms

Exclamation markEnvironment

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

201.2 °F - closed cup

Flash Point (°C)

94 °C - closed cup

PPE (personal protective equipment)

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library