Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

440124

Sigma-Aldrich

Dichlorodiphenylsilane

≥95%

Synonym(s):

Diphenyldichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2SiCl2
CAS Number:
Molecular Weight:
253.20
Beilstein:
609882
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

>1 (vs air)

vapor pressure

2 mmHg ( 125 °C)

Assay

≥95%

form

liquid

refractive index

n20/D 1.578 (lit.)

bp

305 °C (lit.)

density

1.204 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(c1ccccc1)c2ccccc2

InChI

1S/C12H10Cl2Si/c13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

OSXYHAQZDCICNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

287.6 °F - closed cup

Flash Point(C)

142 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Supports for reverse-phase high-performance liquid chromatography of large proteins.
R V Lewis et al.
Analytical biochemistry, 104(1), 153-159 (1980-05-01)
P Huttenloch et al.
Environmental science & technology, 35(21), 4260-4264 (2001-11-23)
The efficacy of the surface modification of natural diatomite and zeolite material by chlorosilanes is demonstrated. Chlorosilanes used were trimethylchlorosilane (TMSCI), tert-butyldimethylchlorosilane (TBDMSCI), dimethyloctadecylchlorosilane (DMODSCI), and diphenyldichlorosilane (DPDSCI) possessing different headgroups and chemical properties. Silanol groups of the diatomite and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service