Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

436186

Sigma-Aldrich

Sodium hexafluoroacetylacetonate

97%

Synonym(s):

Sodium 1,1,1,5,5,5-hexafluoroacetylacetonate, Sodium hexafluoroacetylacetonate, Sodium hexafluoroacetylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaC5HF6O2
CAS Number:
Molecular Weight:
230.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder or crystals

reaction suitability

core: sodium

SMILES string

[Na+].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F

InChI

1S/C5H2F6O2.Na/c6-4(7,8)2(12)1-3(13)5(9,10)11;/h1,12H;/q;+1/p-1/b2-1-;

InChI key

AWUDPEKDPFGDOP-ODZAUARKSA-M

Application

Recently employed in the solid-phase synthesis of chromium β-diketonates, in the preparation of novel catalytic ruthenium diphosphine complexes and in the preparation of a series of new copper chemical vapor deposition (CVD) precursors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rec. Trav. Chim., 114, 242-242 (1995)
Zh. Neorg. Khim., 41, 411-411 (1996)
Organometallics, 14, 2641-2641 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service