Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

433330

Sigma-Aldrich

2-Methylhydrocinnamic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2CH2CO2H
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

Quality Level

Assay

99%

mp

100-103 °C (lit.)

functional group

carboxylic acid

SMILES string

Cc1ccccc1CCC(O)=O

InChI

1S/C10H12O2/c1-8-4-2-3-5-9(8)6-7-10(11)12/h2-5H,6-7H2,1H3,(H,11,12)

InChI key

JIRKNEAMPYVPTD-UHFFFAOYSA-N

General description

2-Methylhydrocinnamic acid is a carboxylic acid building block. It participates in the synthesis of 2-methyl-3-phenylpropanol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Holger Bühler et al.
Chembiochem : a European journal of chemical biology, 6(4), 711-717 (2005-04-07)
The influence of Trp128-substituted mutants of the hydroxynitrile lyase from Manihot esculenta (MeHNL) on the stereoselectivity of MeHNL-catalyzed HCN additions to aldehydes with stereogenic centers, which yield the corresponding cyanohydrins, is described. In rac-2-phenylpropionaldehyde (rac-1) reactions, wild-type (wtMeHNL) and all

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service