429287
(Dimethylamino)trimethyltin(IV)
technical grade
Synonym(s):
Pentamethylstannanamine
About This Item
Recommended Products
grade
technical grade
Quality Level
form
liquid
refractive index
n20/D 1.463 (lit.)
bp
126 °C (lit.)
density
1.274 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CN(C)[Sn](C)(C)C
InChI
1S/C2H6N.3CH3.Sn/c1-3-2;;;;/h1-2H3;3*1H3;/q-1;;;;+1
InChI key
HQFPMGPCIKGRON-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
35.6 °F - closed cup
Flash Point(C)
2 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service