Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4270101

Sigma-Aldrich

Novatach VLVG GRGDSP

GRGDSP-peptide coupled high G low MW sodium alginate, guluronic acid contentguluronic acid content ≥60 %

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

form

powder

composition

guluronic acid content

guluronic acid content, ≥60%

impurities

≤100 EU/g Endotoxins

pH

5.5-8.5 (20 °C, 1% in water)

Application

State-of-the-art, ultra-purified and modified sodium alginate, grafted with RGD peptide sequences, available in 100 mg lyophilized powder vials. Novatach Peptide-coupled UP Alginates are designed to facilitate and promote interaction between cells and alginate-based scaffolds and matrices. They are useful when cell-to-matrix interactions are beneficial. The GRGDSP peptide sequence is a common example of such adhesion ligand used for MSC stem cells. The peptide alginate is available with four different alginate backbones giving flexibility to match hydrogel properties in your application. Examples of Novatach Peptide-coupled UP Alginates applications: initiator for biological interactions between alginate hydrogels and cells, encapsulation of living cells and therapeutic proteins for cell therapy medications, 3D bioprinting, foams and cell tissue scaffolds, hydrogels, advanced drug delivery, etc.

Legal Information

Product of Dupont Nutrition USA, Inc.
NOVATACH is a trademark of Dupont Nutrition USA, Inc.

Disclaimer

NOT FOR USE IN HUMANS

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service