Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

425907

Sigma-Aldrich

Glyceryl tri(palmitate-1-13C)

99 atom % 13C, 98% (CP)

Synonym(s):

1,2,3-propanetriol tris(hexadecanoate-1-13C), Glyceryl tri(hexadecanoate-1-13C), Tripalmitin-carboxyls-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)1413CO2CH2]2CHO213C(CH2)14CH3
CAS Number:
Molecular Weight:
810.30
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

powder

mp

64-66 °C (lit.)

mass shift

M+3

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCC[13C](=O)OCC(CO[13C](=O)CCCCCCCCCCCCCCC)O[13C](=O)CCCCCCCCCCCCCCC

InChI

1S/C51H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h48H,4-47H2,1-3H3/i49+1,50+1,51+1

InChI key

PVNIQBQSYATKKL-SRWOPVBSSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J B Watkins et al.
Gastroenterology, 82(5 Pt 1), 911-917 (1982-05-01)
Three substrates labeled with nonradioactive 13C have been employed to establish a trilogy of noninvasive breath tests to detect fat malabsorption in children and then to differentiate the etiology of the steatorrhea. Administration of 17 mg/kg of (13C)triolein Lipomul (The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service