Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

425745

Sigma-Aldrich

Yttrium(III) trifluoromethanesulfonate

98%

Synonym(s):

Trifluoromethanesulfonic acid yttrium(III) salt, Yttrium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3Y
CAS Number:
Molecular Weight:
536.11
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

core: yttrium
reagent type: catalyst

SMILES string

[Y+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Y/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

JPJIEXKLJOWQQK-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Recently used to promote glycosidation of glycosyl fluorides and as a catalyst in the preparation of pyridine and quinoline derivatives.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 1195-1195 (1995)
Heterocycles, 42, 795-795 (1996)
Synthesis, 801-801 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service