Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

424889

Sigma-Aldrich

Rhodium(II) heptafluorobutyrate dimer

97%

Synonym(s):

Dirhodium heptafluorobutyrate dimer, Dirhodium tetrakis(perfluorobutyrate), Tetrakis(perfluorobutyrato)dirhodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CF3CF2CF2CO2)2Rh]2
CAS Number:
Molecular Weight:
1057.93
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: rhodium

mp

>300 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(=O)O[Rh]OC(=O)C(F)(F)C(F)(F)C(F)(F)F.FC(F)(F)C(F)(F)C(F)(F)C(=O)O[Rh]OC(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/4C4HF7O2.2Rh/c4*5-2(6,1(12)13)3(7,8)4(9,10)11;;/h4*(H,12,13);;/q;;;;2*+2/p-4

InChI key

BNCXPORMMOBRMR-UHFFFAOYSA-J

Application

Recently used in the catalytic preparation of macrocycles and in the efficient synthesis of chiral β-substituted (E)-crotylsilanes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 36, 8723-8723 (1996)
Journal of the American Chemical Society, 118, 7865-7865 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service