Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

422916

Sigma-Aldrich

2-Methoxy-4-nitrobenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
197.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

146-148 °C (lit.)

functional group

carboxylic acid
nitro

SMILES string

COc1cc(ccc1C(O)=O)[N+]([O-])=O

InChI

1S/C8H7NO5/c1-14-7-4-5(9(12)13)2-3-6(7)8(10)11/h2-4H,1H3,(H,10,11)

InChI key

KPJXEWJRJKEOCD-UHFFFAOYSA-N

General description

2-Methoxy-4-nitrobenzoic acid is an alkoxybenzoic acid derivative. It has been reported to be synthesized by reacting 2-hydroxy-4-nitrobenzoic acid with methyl iodide and characterized by 1H and 13C-NMR spectra.

Application

2-Methoxy-4-nitrobenzoic acid may be used as a starting material in the following syntheses:
  • 2-Methoxy-4-nitrobenzamide, a nitroamide derivative.
  • 4-Amino-2-methoxybenzamide, an aminobenzamide derivative.
  • N,2-Dimethoxy-N-methyl-4-nitrobenzamide, a Weinreb amide derivative.
  • 1-(2-Methoxy-4-nitrophenyl)-5-methylhex-2-yn-1-one, a ynone derivative.
  • 2,5-Constrained piperidine derivative, a potential CCR3 (Chemokine (C-C Motif) Receptor 3) antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leyi Gong et al.
Bioorganic & medicinal chemistry letters, 13(20), 3597-3600 (2003-09-25)
As part of our investigation into the development of potent CCR3 antagonists, a series of piperidine analogues was designed and prepared. Exploration of the piperidine core examined both the basicity and the location of a nitrogen, as well as conformational
Marc J Adler et al.
The Journal of organic chemistry, 76(17), 7040-7047 (2011-07-12)
The design and synthesis of small molecule α-helix mimetics has been a productive field over the past decade. These compounds have performed well in a variety of biological systems as functional disruptors of α-helix-mediated protein-protein interactions. In our studies we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service