Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

421073

Sigma-Aldrich

TYZOR® 131 organic titanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.22

vapor pressure

583 mmHg ( 100 °C)

reaction suitability

core: titanium
reagent type: catalyst

refractive index

n20/D 1.4120 (lit.)

density

1.265 g/mL at 25 °C (lit.)

Legal Information

TYZOR is a registered trademark of E. I. du Pont de Nemours and Company

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service