Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

418420

Sigma-Aldrich

Poly(vinyl butyral-co-vinyl alcohol-co-vinyl acetate)

average Mw 170,000-250,000, powder

Synonym(s):

Poly(vinyl butyral-vinyl alcohol-vinyl acetate)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mw 170,000-250,000

composition

acetate, 0-2.5 wt. %
hydroxyl, 17.5-20 wt. %
vinyl butyral, 80 wt. %

impurities

<3.5 wt. % Volatiles

viscosity

170-260 cSt, 7.5 wt. % in anhydrous methanol(20 °C, Ostwald)(lit.)

transition temp

Tg 72-78 °C

solubility

alcohols, glycol ethers and dioxane: soluble

density

1.083 g/mL at 25 °C (lit.)

InChI

1S/C8H14O2.C4H6O2.C2H4O/c1-4-7-8(9-5-2)10-6-3;1-3-6-4(2)5;1-2-3/h5-6,8H,2-4,7H2,1H3;3H,1H2,2H3;2-3H,1H2

InChI key

RNNFCYZWIQZWQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ceramic binder, dry toners, wood coatings, composite fiber binders, structural adhesives and interlayer in safety glass.

Features and Benefits

Good binding efficiency, optical clarity and adhesion to a variety of surfaces.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service