Skip to Content
MilliporeSigma
All Photos(1)

Documents

416789

Sigma-Aldrich

1,1,1-Trifluoro-3-phenyl-2-propanone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2COCF3
CAS Number:
Molecular Weight:
188.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.444 (lit.)

bp

51-52 °C/2 mmHg (lit.)

density

1.22 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(=O)Cc1ccccc1

InChI

1S/C9H7F3O/c10-9(11,12)8(13)6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

IAJKTOIWQHTZOS-UHFFFAOYSA-N

Related Categories

General description

1,1,1-Trifluoro-3-phenyl-2-propanone is a trifluoromethyl ketone. Its in vitro neuroprotective potency against low K(+)-induced apoptosis in cerebellar granule neurons (CGNs) by different pathways has been reported.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katsuyoshi Sunaga et al.
In vivo (Athens, Greece), 16(2), 97-101 (2002-06-21)
We previously reported that two trifluoromethyl ketones, 3,3,3-trifluoro-1-phenyl-1,2-propanedione (TF1) and 1,1,1-trifluoro-3-phenyl-2-propanone (TF2), have neuroprotective effects against low K(+)-induced apoptosis in cerebellar granule neurons (CGNs) exposed at 12-13 days in vitro (DIV). On the other hand, these compounds showed weak neuroprotective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service