Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

416452

Sigma-Aldrich

Dimethyl sulfide-d6

99 atom % D

Synonym(s):

(Methyl sulfide)-d6, Hexadeuterodimethyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3SCD3
CAS Number:
Molecular Weight:
68.17
Beilstein:
2036892
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99 atom % D

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.431 (lit.)

bp

36.5 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])SC([2H])([2H])[2H]

InChI

1S/C2H6S/c1-3-2/h1-2H3/i1D3,2D3

InChI key

QMMFVYPAHWMCMS-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfide-d6 [(CD3)2S] is a deuterated NMR solvent useful in NMR-based research and analyses. It can form 1:1 complex with halothane by the formation of C–H…S hydrogen bonds. During the reaction of (CD3)2S with singlet oxygen in aprotic solvents, the H-D exchange in the methyl group was observed. The electronic spectra of (CD3)2S has been measured between 1250 and 2500Å.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-34.6 °F - closed cup

Flash Point(C)

-37 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Vibrational spectra of (CH3)2S, CH3SCD3, and (CD3)2S.
Ypenburg JW and Gerding H.
Rec. Trav. Chim., 90(8), 885-895 (1971)
Assignments of Rydberg and valence transitions in the electronic absorption spectrum of dimethyl sulfide.
McDiarmid R.
J. Chem. Phys. , 61(1), 274-281 (1974)
Intramolecular potential function and methyl-methyl coupling of crystalline dimethyl sulfide.
Levin IW, et al.
Spectrochimica Acta Part A: Molecular Spectroscopy, 31(1), 41-55 (1975)
C-H?X (X= S, P) hydrogen bonding: The complexes of halothane with dimethyl sulfide and trimethylphosphine.
Michielsen B, et al.
Journal of Molecular Structure, 1023, 90-95 (2012)
Mechanism of sulfone formation in the reaction of sulfides and singlet oxygen: intermediacy of S-hydroperoxysulfonium ylide.
Ishiguro K, et al.
Journal of the American Chemical Society, 118(31), 7265-7271 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service