Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

412783

Sigma-Aldrich

Dimethyl 1,1-cyclopropanedicarboxylate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H4(CO2CH3)2
CAS Number:
Molecular Weight:
158.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

196-198 °C (lit.)

density

1.147 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)C1(CC1)C(=O)OC

InChI

1S/C7H10O4/c1-10-5(8)7(3-4-7)6(9)11-2/h3-4H2,1-2H3

InChI key

PWLLZZMFFZUSOG-UHFFFAOYSA-N

Application

Dimethyl 1,1-cyclopropanedicarboxylate may be used in the preparation of α-diazo-β-ketoester.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric approach to the pentacyclic skeleton of Aspidosperma alkaloids via enantioselective intramolecular 1, 3-dipolar cycloaddition of carbonyl ylides catalyzed by chiral dirhodium (II) carboxylates.
Nambu H, et al.
Tetrahedron Letters, 50(26), 3675-3678 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service