Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

411310

Sigma-Aldrich

(Carbethoxymethyl-1,2-13C2)triphenylphosphonium bromide

98 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5O213C13CH2P(C6H5)3Br
CAS Number:
Molecular Weight:
431.27
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % 13C

form

solid

mol wt

431.23 by atom % calculation

reaction suitability

reaction type: C-C Bond Formation
reaction type: Whiting Reaction

mp

152-154 °C (lit.)

mass shift

M+2

SMILES string

[Br-].CCO[13C](=O)[13CH2][P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C22H22O2P.BrH/c1-2-24-22(23)18-25(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21;/h3-17H,2,18H2,1H3;1H/q+1;/p-1/i18+1,22+1;

InChI key

VJVZPTPOYCJFNI-WWPGYFPTSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service