Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

407534

Sigma-Aldrich

Tris[2-(acryloyloxy)ethyl] isocyanurate

Synonym(s):

Tri(2-hydroxyethyl)isocyanurate triacrylate, Tris(2-hydroxyethyl) isocyanurate triacrylate, Tris(2-hydroxyethyl)isocyanuric acid triacrylate, Tris[2-(acryloyloxy)ethyl]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N3O9
CAS Number:
Molecular Weight:
423.37
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

contains

100 ppm monomethyl ether hydroquinone as inhibitor

mp

52-53 °C (lit.)

SMILES string

C=CC(=O)OCCN1C(=O)N(CCOC(=O)C=C)C(=O)N(CCOC(=O)C=C)C1=O

InChI

1S/C18H21N3O9/c1-4-13(22)28-10-7-19-16(25)20(8-11-29-14(23)5-2)18(27)21(17(19)26)9-12-30-15(24)6-3/h4-6H,1-3,7-12H2

InChI key

YIJYFLXQHDOQGW-UHFFFAOYSA-N

General description

Tris[2-(acryloyloxy)ethyl] isocyanurate is a compound belonging to the class of acrylate polymers, recognized for its multifunctional capabilities and applications in various fields, particularly in biomedicine and materials science. In the biomedical field, Tris[2-(acryloyloxy)ethyl] isocyanurate is primarily utilized in dental materials, tissue engineering, and as a crosslinking agent in polymer formulations. In dental applications, it serves as a key component in dental adhesives and composites, enhancing the mechanical properties and durability of restorative materials. Additionally, its ability to form hydrogels makes it suitable for drug delivery systems, where it can facilitate controlled release of therapeutic agents, and for tissue engineering scaffolds, promoting cell adhesion and proliferation due to its biocompatibility.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caitlyn C Cook et al.
Advanced materials (Deerfield Beach, Fla.), 32(47), e2003376-e2003376 (2020-10-02)
Volumetric additive manufacturing (VAM) forms complete 3D objects in a single photocuring operation without layering defects, enabling 3D printed polymer parts with mechanical properties similar to their bulk material counterparts. This study presents the first report of VAM-printed thiol-ene resins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service