Skip to Content
MilliporeSigma

406554

Diethyl malonimidate dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C2H5OC(=NH)CH2C(=NH)OC2H5·2HCl
CAS Number:
Molecular Weight:
231.12
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

impurities

<5% ammonium chloride

mp

122 °C (dec.) (lit.)

functional group

amine, ether

SMILES string

Cl.Cl.CCOC(=N)CC(=N)OCC

InChI

1S/C7H14N2O2.2ClH/c1-3-10-6(8)5-7(9)11-4-2;;/h8-9H,3-5H2,1-2H3;2*1H

InChI key

TWUQXVGHXWRUBR-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of rhenium cyanobis(oxazoline) oxo complexes as enantioselective reduction catalysts
  • Synthesis of chiral bis(oxazoline)-copper complexes immobilized by donor-acceptor interactions on insoluble organic supports as reusable catalysts for asymmetric Diels-Alder cycloaddition
  • Preparation of nonracemic bis(oxazoline)ruthenium p-cymene complexes and silica-supported analogs as catalysts for the enantioselective transfer hydrogenation of ketones to secondary alcohols
Diethyl malonimidate dihydrochloride may be used in the preparation of chiral bis(oxazoline) ligands. It may be used in the preparation of intramolecularly cross-linked urokinase.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library