Skip to Content
MilliporeSigma
All Photos(2)

Documents

404950

Sigma-Aldrich

Titanium(IV) methoxide

95%

Synonym(s):

Methyl orthotitanate, Tetramethoxytitanium, Tetramethyl orthotitanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti(OCH3)4
CAS Number:
Molecular Weight:
172.00
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid
solid

reaction suitability

core: titanium
reagent type: catalyst

impurities

<5% methanol

mp

200-210 °C (lit.)

SMILES string

CO[Ti](OC)(OC)OC

InChI

1S/4CH3O.Ti/c4*1-2;/h4*1H3;/q4*-1;+4

InChI key

ITNVWQNWHXEMNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Used for the preparation of polyoxotitanates.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service