Skip to Content
MilliporeSigma
All Photos(2)

Documents

399019

Sigma-Aldrich

6′-Methoxy-2′-acetonaphthone

98%

Synonym(s):

1-(6-Methoxy-2-naphthalenyl)ethanone, 2-Acetyl-6-methoxynaphthalene, NSC 105564

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6COCH3
CAS Number:
Molecular Weight:
200.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

107-109 °C (lit.)

SMILES string

COc1ccc2cc(ccc2c1)C(C)=O

InChI

1S/C13H12O2/c1-9(14)10-3-4-12-8-13(15-2)6-5-11(12)7-10/h3-8H,1-2H3

InChI key

GGWCZBGAIGGTDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6′-Methoxy-2′-acetonaphthone (6-methoxy-2-naphthylacetic acid ) is metabolite of nabumetone, a phototoxic nonsteroidal antiinflammatory drug.1
6′-Methoxy-2′-acetonaphthone is suitable for use in the synthesis of 6-methoxy-2-naphthylacetic acid. It may be used in the preparation of fluorogenic aldol sensors.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Boscá et al.
Photochemistry and photobiology, 71(2), 173-177 (2000-02-25)
Nabumetone is a phototoxic nonsteroidal antiinflammatory drug used for the treatment of osteoarthritis. However, nabumetone is considered a prodrug with its metabolite 6-methoxy-2-naphthylacetic acid the active form. Photophysical and photochemical studies on this metabolite have been undertaken. It undergoes photodecarboxylation
M Nobilis et al.
Journal of pharmaceutical and biomedical analysis, 32(4-5), 641-656 (2003-08-06)
The disposition of the non-steroidal anti-inflammatory drug (NSAID) nabumetone after a single oral dose administration of nabumetone tablets to humans and minipigs was investigated. Nabumetone is a prodrug, which is metabolized in the organism to the principal pharmacodynamically active metabolite
B List et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(26), 15351-15355 (1998-12-23)
The synthesis of novel fluorogenic retro-aldol substrates for aldolase antibody 38C2 is described. These substrates are efficiently and specifically processed by antibody aldolases but not by natural cellular enzymes. Together, the fluorogenic substrates and antibody aldolases provide reporter gene systems

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service