Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

396613

Mercury absorption kit

size 6 1/2 in. × 3 3/4 in. × 2 1/4 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131905
NACRES:
NB.35

size

6 1/2 in. × 3 3/4 in. × 2 1/4 in.

General description

Mercurisorb has been proven superior to iodine, coal, powdered zinc, and sulfur blossom for absorbing mercury. Absorbs its own weight in less than five minutes for easy disposal. Kit includes: 100g bottle of Mercurisorb, brush and minishovel, 3 pairs of disposable gloves, container for absorption mixture, disposable pipettes, and a compact PS case.

Legal Information

Mercurisorb is a trademark of Carl Roth GmbH & Co.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service