Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

395005

Supelco

N,N-Dimethylformamide di-tert-butyl acetal

for GC derivatization, LiChropur

Synonym(s):

1,1-Di-tert-butoxy-N,N-dimethylmethylamine, 1,1-Di-tert-butoxytrimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH[OC(CH3)3]2
CAS Number:
Molecular Weight:
203.32
Beilstein:
969629
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Assay

≥90.0%

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.413 (lit.)

bp

56-57 °C/8 mmHg (lit.)

density

0.848 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(OC(C)(C)C)OC(C)(C)C

InChI

1S/C11H25NO2/c1-10(2,3)13-9(12(7)8)14-11(4,5)6/h9H,1-8H3

InChI key

DBNQIOANXZVWIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylformamide di-tert-butyl acetal is an derivatizing reagent.

Application

It was used as derivatizing agent in a study to determine cocaine and benzoyl ecgonine in urine using GC with on-column alkylation.
Reagent used for the preparation of indolizines via intermolecular cyclization of picolinium salts.

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 8817-8817 (2006)
N C Jain et al.
Journal of forensic sciences, 22(1), 7-16 (1977-01-01)
A gas chromatographic procedure has been developed for the simultaneous determination of cocaine and benzoyl ecgonine in urine specimens. The two drugs are extracted by isopropanol/chloroform from urine samples saturated with a bisalt buffer. The organic extract is evaporated to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service