Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

394521

Sigma-Aldrich

3-Bromophenethyl alcohol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2CH2OH
CAS Number:
Molecular Weight:
201.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
99%

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.5732 (lit.)

bp

107-110 °C/1 mmHg (lit.)

density

1.478 g/mL at 25 °C (lit.)

functional group

bromo
hydroxyl

SMILES string

OCCc1cccc(Br)c1

InChI

1S/C8H9BrO/c9-8-3-1-2-7(6-8)4-5-10/h1-3,6,10H,4-5H2

InChI key

PTTFLKHCSZSFOL-UHFFFAOYSA-N

General description

3-Bromophenethyl alcohol is a phenethyl alcohol derivative. Its enthalpy of vaporization at boiling point has been determined.

Application

3-Bromophenethyl alcohol may be used as a starting material in the synthesis of its mesylate derivative (m-Br-C6H4CH2CH2OMs) by treatment with mesyl chloride.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annie Tam et al.
Bioorganic & medicinal chemistry, 17(3), 1055-1063 (2008-03-04)
The traceless Staudinger ligation can be mediated by phosphinothiols under physiological conditions. Proximal positive charges are necessary to achieve that transformation, presumably because those charges discourage protonation of the key iminophosphorane intermediate. Here, a series of cationic phosphinothiols is used
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 466-466 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service