Skip to Content
MilliporeSigma
All Photos(1)

Documents

393983

Sigma-Aldrich

Benzophenone-2,3,4,5,6-d5

98 atom % D

Synonym(s):

2,3,4,5,6-Pentadeuterobenzophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COC6D5
CAS Number:
Molecular Weight:
187.25
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Quality Level

form

solid

bp

305 °C (lit.)

mp

47-51 °C (lit.)

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(=O)c2ccccc2

InChI

1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H/i1D,3D,4D,7D,8D

InChI key

RWCCWEUUXYIKHB-DYVTXVBDSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristina Kotnik et al.
Chemosphere, 147, 114-123 (2016-01-15)
This study investigates the environmental fate of eight benzophenone derivatives (the pharmaceutical ketoprofen, its phototransformation products 3-ethylbenzophenone and 3-acetylbenzophenone, and five benzophenone-type UV filters) by evaluating their photolytic behaviour. In addition, the genotoxicity of these compounds and the produced photodegradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service