All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H10O
CAS Number:
Molecular Weight:
146.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.558 (lit.)
bp
70-72 °C/0.7 mmHg (lit.)
density
1.075 g/mL at 25 °C (lit.)
functional group
ketone
SMILES string
CC1CC(=O)c2ccccc12
InChI
1S/C10H10O/c1-7-6-10(11)9-5-3-2-4-8(7)9/h2-5,7H,6H2,1H3
InChI key
XVTQSYKCADSUHN-UHFFFAOYSA-N
General description
3-Methyl-1-indanone is a derivative of 1-indanone.Its synthesis has been reported. The 1H and 13C-NMR spectra of 3-methyl-1-indanone has been reported. Biocatalyzed oxidation of racemic 3-methyl-1-indanone with high enanatioselectivity has been investigated.
Application
3-Methyl-1-indanone is suitable for the synthesis of branched alkyl indanes (BINs). It may be used in the following studies:
- Synthesis of 2,2,3-tribromo-2,3-dihydro-3-methylinden-1-one and 2-bromo-3-bromomethyl-1H-inden-1-one.
- As photosensitizers for the splitting of dimethylthymine dimers.
- As a model to verify three-phase model in enantioselective gas-liquid chromatography.
- (-)-(R)-3-methyl-1-indanone may be used as standard for comparing the optical rotatory dispersion curve with 1,3-dialkylindenes in the study of base-catalyzed 1,3 proton transfer in indene systems.
- As an optically active sensitizer to induce asymmetry in trans-1,2-diphenylcylcopropane (DPC).
- The dl-isomers of 3-methyl-1-indanone may be used in the synthesis of monomeric and trimeric 3-methyl-1-indanethione.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thiocarbonyls. VI. Studies in the Indanone and Tetralone Series1.
Campaigne E and Moss RD.
Journal of the American Chemical Society, 76(5), 1269-1271 (1954)
Asymmetric induction during transfer of triplet energy.
Ouannes C, et al.
Journal of the American Chemical Society, 95(25), 8472-8474 (1973)
Ana Rioz-Martínez et al.
The Journal of organic chemistry, 75(6), 2073-2076 (2010-02-20)
Baeyer-Villiger monooxygenases have been tested in the oxidation of racemic benzofused ketones. When employing a single mutant of phenylacetone monooxygenase (M446G PAMO) under the proper reaction conditions, it was possible to achieve 3-substituted 3,4-dihydroisocoumarins with high yields and optical purities
Electrophilic substitution at saturated carbon. XLIII. Alkylammonium carbanide ion-pair reorganization reactions in base-catalyzed 1,3-proton transfer in an indene system.
Almy J and Cram DJ.
Journal of the American Chemical Society, 91(16), 4459-4468 (1969)
Photochemical bromination of substituted indan-1-one derivatives: synthesis of new polybromoindan-1-one derivatives.
KUS, NS.
Turkish Journal of Chemistry, 33(4), 479-486 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service