Skip to Content
MilliporeSigma
All Photos(1)

Documents

382434

Sigma-Aldrich

(S)-(−)-1,1′-Binaphthyl-2,2′-diamine

99%

Synonym(s):

(S)-(−)-1,1′-Bi(2-naphthylamine), (S)-(−)-1,1′-Binaphthalene-2,2′-diamine, (S)-(−)-2,2′-Diamino-1,1′-binaphthalene, (S)-(−)-DABN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H6C10H6NH2
CAS Number:
Molecular Weight:
284.35
Beilstein:
2419071
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

optical activity

[α]20/D −157°, c = 1 in pyridine

optical purity

ee: 99% (HPLC)

mp

242-244 °C (lit.)

SMILES string

Nc1ccc2ccccc2c1-c3c(N)ccc4ccccc34

InChI

1S/C20H16N2/c21-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)22/h1-12H,21-22H2

InChI key

DDAPSNKEOHDLKB-UHFFFAOYSA-N

Application

(S)-(-)-1,1′-Binaphthyl-2,2′-diamine may be used in the synthesis of (S)-(+)-N,N′-dimethyl-N,N′-bis{3-[bis-(1-methyl-2-benzimidazolyl methyl)]-amino]-propyl}-1,1′-binaphthyl-2,2′-diamine, an octadentate ligand that can form dinuclear and trinuclear copper(II) complexes. It may also be used in the synthesis of (Sa)-N-[2´-amino-(1,1´-binaphthyl)-2-yl]-4-methylbenzenesulfonamide.
Used in the synthesis of chiral lactones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Models for biological trinuclear copper clusters. Characterization and enantioselective catalytic oxidation of catechols by the copper (II) complexes of a chiral ligand derived from (S)-(-)-1, 1'-binaphthyl-2, 2'-diamine.
Mimmi MC, et al.
Dalton Transactions, 14, 2192-2201 (2004)
The Journal of Organic Chemistry, 56, 1112-1112 (1991)
(Sa, S)-N-[2'-(4-Methylphenylsulfonamido)-1, 1'-Binaphthyl-2-Yl] Pyrrolidine-2-Carboxamide: An Organocatalyst for the Direct Aldol Reaction.
Vizoquez SF, et al.
Organic Syntheses, 317-329 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service