Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

380741

Sigma-Aldrich

1,1,1-Tris(diphenylphosphinomethyl)ethane

Synonym(s):

1,3-Bis(diphenylphosphino)-2-(diphenylphosphino)methyl-2-methylpropane, Triphos

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C[CH2P(C6H5)2]3
CAS Number:
Molecular Weight:
624.67
Beilstein:
967586
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

99-102 °C (lit.)

functional group

phosphine

SMILES string

CC(CP(c1ccccc1)c2ccccc2)(CP(c3ccccc3)c4ccccc4)CP(c5ccccc5)c6ccccc6

InChI

1S/C41H39P3/c1-41(32-42(35-20-8-2-9-21-35)36-22-10-3-11-23-36,33-43(37-24-12-4-13-25-37)38-26-14-5-15-27-38)34-44(39-28-16-6-17-29-39)40-30-18-7-19-31-40/h2-31H,32-34H2,1H3

InChI key

BARUNXKDFNLHEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organometallics, 9, 226-226 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service