Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

380423

Sigma-Aldrich

Triethylsilanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3SiOH
CAS Number:
Molecular Weight:
132.28
Beilstein:
1732848
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.433 (lit.)

bp

158 °C (lit.)
86-87 °C/33 mmHg (lit.)

density

0.864 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC[Si](O)(CC)CC

InChI

1S/C6H16OSi/c1-4-8(7,5-2)6-3/h7H,4-6H2,1-3H3

InChI key

WVMSIBFANXCZKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika Stróżyńska et al.
Journal of chromatography. A, 1597, 1-8 (2019-03-18)
A new, simple derivatization method for the determination of perfluorocarboxylic acids (PFCAs, C3-C12) in water is presented. The method is based on an acid-catalyzed esterification of perfluorinated carboxylic acids with triethylsilanol. During the reaction, a triethylsilyl ester is formed which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service