Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

379239

Sigma-Aldrich

Trimethylaluminum solution

2.0 M in heptane

Synonym(s):

Aluminum trimethanide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3Al
CAS Number:
Molecular Weight:
72.09
Beilstein:
3587197
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

2.0 M in heptane

density

0.693 g/mL at 25 °C

SMILES string

C[Al](C)C

InChI

1S/3CH3.Al/h3*1H3;

InChI key

JLTRXTDYQLMHGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethylaluminum can be used in the synthesis of methylaluminoxane (MAO), a precatalyst for olefin polymerization. It is a precursor for the preparation of a wide range of organoaluminium such as dimethylaluminum chloride and bis(trimethylaluminum)-1,4-diazabicyclo[2.2.2]-octane (DABAL-Me3).

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Remarkably Stable (Me3Al) 2? DABCO and Stereoselective Nickel?Catalyzed AlR3 (R= Me, Et) Additions to Aldehydes.
Biswas K, et al.
Angewandte Chemie (International Edition in English), 44(15), 2232-2234 (2005)
Gasometric titration for dimethylaluminum chloride analysis.
Wang L, et al.
Journal of Pharmaceutical and Biomedical Analysis, 125, 110-113 (2016)
Methylaluminoxane: synthesis, characterization and catalysis of ethylene polymerization.
Reddy S S, et al.
Polymer Bull., 36(2), 165-171 (1996)
J L L Robinson et al.
Oncogene, 33(50), 5666-5674 (2013-12-03)
Castration-resistant prostate cancer (CRPC) continues to pose a significant clinical challenge with new generation second-line hormonal therapies affording limited improvement in disease outcome. As the androgen receptor (AR) remains a critical driver in CRPC, understanding the determinants of its transcriptional
Shahram Khosravi et al.
The Journal of investigative dermatology, 135(5), 1358-1367 (2015-01-07)
Human cutaneous melanoma is a devastating skin cancer because of its invasive nature and high metastatic potential. We used tissue microarray to study the role of human eukaryotic translation initiation factor 4E (eIF4E) in melanoma progression in 448 melanocytic lesions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service