Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

377937

Sigma-Aldrich

3-Nitrophenylacetonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2CN
CAS Number:
Molecular Weight:
162.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

180 °C/3 mmHg (lit.)

mp

60-62 °C (lit.)

functional group

nitrile
nitro

SMILES string

[O-][N+](=O)c1cccc(CC#N)c1

InChI

1S/C8H6N2O2/c9-5-4-7-2-1-3-8(6-7)10(11)12/h1-3,6H,4H2

InChI key

WAVKEPUFQMUGBP-UHFFFAOYSA-N

General description

3-Nitrophenylacetonitrile is a builing block. Reactions of 3-nitrophenylacetonitrile with isopropyl alcohol, cyclohexyl alcohol and benzyl alcohol at high temperature have been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon-carbon bond cleavage and esterification of phenylacetonitrile and its derivatives affording the corresponding benzoic esters.
Wang Z, et al.
J. Chem. Res. (M), 2007(6), 323-324 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service