Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

370207

Sigma-Aldrich

4,6-Dimethyl-α-pyrone

98%

Synonym(s):

4,6-Dimethyl-2H-pyran-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

48-50 °C (lit.)

functional group

ester

SMILES string

CC1=CC(C)=CC(=O)O1

InChI

1S/C7H8O2/c1-5-3-6(2)9-7(8)4-5/h3-4H,1-2H3

InChI key

IXYLIUKQQQXXON-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

General description

4,6-Dimethyl-α-pyrone was immobilized in a guanidinium-sulfonate-calixarene (G4C) crystalline network, which upon exposure to ultraviolet irradiation, transforms the entrapped 4,6-dimethyl-α-pyrone into a 4,6-dimethyl-β-lactone. Thin solid films layers of 4,6-dimethyl-α-pyrone on UV irradiation at -190°C were investigated.

Application

4,6-Dimethyl-α-pyrone (Me21) has been used in the preparation of G4C{Me21} via reacting with tetra-p-sulfocalix[4]arene (C) and guanidine hydrochloride (GCl). It may be used in the preparation of host-guest complex G4C{Me21} by reacting with crystalline guanidinium-sulfonate-calixarene (G4C). This complex on UV irradiation afforded 1,3-dimethylcyclobutadiene (Me2CBD).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yves-Marie Legrand et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(36), 10021-10028 (2011-09-08)
Cyclobutadiene (CBD), the smallest cyclic hydrocarbon bearing conjugated double bonds, has long intrigued chemists because of its chemical characteristics. The question of whether the molecule could be prepared at all has been answered, but the parent compound and its unperturbed
Yves-Marie Legrand et al.
Science (New York, N.Y.), 329(5989), 299-302 (2010-07-22)
Cyclobutadiene (CBD), the smallest cyclic hydrocarbon bearing conjugated double bonds, has long intrigued chemists on account of its strained geometry and electronic instability, but the parent compound and its unperturbed derivatives have thus far eluded crystallographic characterization. In this work
4, 6-Dimethyl-a-pyrone: a matrix isolation study of the photochemical generation of conjugated ketene, Dewar valence isomer and 1, 3-dimethyl-cyclobutadiene.
Breda S, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 162(1), 139-151 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service