Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

366943

Sigma-Aldrich

(S)-(−)-1-Benzyl-3-pyrrolidinol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO
CAS Number:
Molecular Weight:
177.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

optical activity

[α]24/D −3.7°, c = 5 in methanol

refractive index

n20/D 1.548 (lit.)

bp

115 °C/0.8 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O[C@H]1CCN(C1)Cc2ccccc2

InChI

1S/C11H15NO/c13-11-6-7-12(9-11)8-10-4-2-1-3-5-10/h1-5,11,13H,6-9H2/t11-/m0/s1

InChI key

YQMXOIAIYXXXEE-NSHDSACASA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service