Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

364096

Sigma-Aldrich

1-Aza-15-crown-5

97%

Synonym(s):

1,4,7,10-Tetraoxa-13-azacyclopentadecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H21NO4
CAS Number:
Molecular Weight:
219.28
Beilstein:
971959
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

35-37 °C (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

C1COCCOCCOCCOCCN1

InChI

1S/C10H21NO4/c1-3-12-5-7-14-9-10-15-8-6-13-4-2-11-1/h11H,1-10H2

InChI key

BJUOQSZSDIHZNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moutusi Banerjee et al.
Small (Weinheim an der Bergstrasse, Germany), 11(28), 3451-3457 (2015-03-12)
Attachment of Li(+) ion on graphene surface to realize Li(+)-ion conductor is a real challenge because of the weak interaction between the ions and the functional groups of graphene oxide; although, a large number of theoretical results are already available

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service