Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

358940

Sigma-Aldrich

6-Heptenoic acid

99%

Synonym(s):

6-Heptanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)4COOH
CAS Number:
Molecular Weight:
128.17
Beilstein:
1747265
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

222-224 °C (lit.)

mp

−6.5 °C (lit.)

density

0.946 g/mL at 25 °C (lit.)

functional group

allyl
carboxylic acid

storage temp.

2-8°C

SMILES string

OC(=O)CCCCC=C

InChI

1S/C7H12O2/c1-2-3-4-5-6-7(8)9/h2H,1,3-6H2,(H,8,9)

InChI key

RWNJOXUVHRXHSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Heptenoic acid is a straight-chain terminal alkenoic acid. Electrochemical oxidation of 6-heptenoic acid adsorbed on Pt(111) electrode surface has been reported. Preparation of 6-heptenoic acid has been reported.

Application

6-Heptenoic acid may be used in the preparation of 6-(iodomethyl)-hexanolide, via iodo-lactonization.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of seven-membered and medium-ring lactones by iodo lactonization.
Simonot B and Rousseau G.
The Journal of Organic Chemistry, 58(1), 4-5 (1993)
1, 2, 6-Tribromohexane, 5-Hexenylmagnesium Bromide, and 6-Heptenoic Acid1.
Lamb RC and Ayers PW.
The Journal of Organic Chemistry, 27(4), 1441-1442 (1962)
Electrochemical oxidation of adsorbed alkenoic acids as a function of chain length at Pt (111) electrodes. Studies by cyclic voltammetry, EELS and Auger spectroscopy.
Batina N, et al.
Catalysis Letters, 3(4), 275-297 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service