Skip to Content
MilliporeSigma
All Photos(1)

Documents

357464

Sigma-Aldrich

Yttrium barium copper oxide

powder, ≤5 μm particle size, 99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YBa2Cu3Ox, x ~ 6.7-9.5
CAS Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9%

form

powder

particle size

≤5 μm

SMILES string

O=[Cu].O=[Ba].O=[Y]O[Y]=O

InChI

1S/Ba.Cu.5O.2Y

InChI key

OHBOMLDLKVPQKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

produced for optimum phase purity (>99%) and sinterability.

Physical form

High Tc superconductor powder

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Krakow
Journal of electron microscopy technique, 18(3), 277-290 (1991-07-01)
A new method has been developed for simulating high resolution electron microscope images of weak phase objects via a digital television frame store system and fast Fourier transforms of a graphical representation of structures containing several atomic species. Here masks
Zhonghua Zhu et al.
Physical chemistry chemical physics : PCCP, 14(11), 3859-3863 (2012-02-14)
Nanoparticles of superconducting YBa(2)Cu(3)O(7-δ) were synthesized via a citrate pyrolysis technique. Room temperature ferromagnetism was revealed in the samples by a vibrating sample magnetometer. Electron spin resonance spectra at selected temperatures indicated that there is a transition from the normal
Kartik Venkataraman et al.
Applied spectroscopy, 59(5), 639-649 (2005-06-23)
Thin film embodiments of MBa2Cu3O7-x (MBCO, M = yttrium or a rare-earth metal) prepared by several different deposition methods on a variety of substrates were investigated by Raman microspectroscopy. Several of the unique characterization capabilities of Raman spectroscopy in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service