Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

348961

Sigma-Aldrich

Oxacyclotridecan-2-one

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.472 (lit.)

bp

150-151 °C/23 mmHg (lit.)

mp

2-3 °C (lit.)

density

0.981 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

O=C1CCCCCCCCCCCO1

InChI

1S/C12H22O2/c13-12-10-8-6-4-2-1-3-5-7-9-11-14-12/h1-11H2

InChI key

DQGSJTVMODPFBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxacyclotridecan-2-one is a lactone (lauryl lactone). It is formed by the Baeyer-Villiger oxidation of cyclododecanone.

Application

Oxacyclotridecan-2-one may be used in the synthesis of fatty acid derivative, (E)-11-methyl-12-octadecenoic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kostichka et al.
Journal of bacteriology, 183(21), 6478-6486 (2001-10-10)
Biological oxidation of cyclic ketones normally results in formation of the corresponding dicarboxylic acids, which are further metabolized in the cell. Rhodococcus ruber strain SC1 was isolated from an industrial wastewater bioreactor that was able to utilize cyclododecanone as the
N M Carballeira et al.
Lipids, 33(6), 627-632 (1998-07-09)
The fatty acid composition of a Pseudomonas sp. (Alteromonas) and its host, the dinoflagellate Ostreopsis lenticularis, vectors in ciguatera fish poisoning, has been studied. The major fatty acids in O. lenticularis were 16:0, 20:5n-3, and 22:6n-3, but 18:2n-6, 18:3n-3, and
Coraline Simon et al.
Toxicology in vitro : an international journal published in association with BIBRA, 37, 121-133 (2016-10-19)
Endocrine activity of 65 compounds migrating from polycarbonate replacement plastic baby bottles was assessed using in vitro cell based assays (reporter gene assays) involving 7 nuclear receptors, i.e. human steroid hormones receptors (oestrogen, androgen, progesterone and glucocorticoid receptors), human thyroid
Matthias Onghena et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 31(12), 2090-2102 (2014-11-20)
In 2011, the European Union prohibited the production of polycarbonate (PC) baby bottles due to the toxic effects of the PC monomer bisphenol-A. Therefore, baby bottles made of alternative materials, e.g. polypropylene (PP) or polyethersulphone (PES), are currently marketed. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service