All Photos(1)
About This Item
Linear Formula:
CH3C(OCH3)2COCH3
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.407 (lit.)
bp
145-146 °C (lit.)
density
0.987 g/mL at 25 °C (lit.)
functional group
ether
ketal
ketone
SMILES string
COC(C)(OC)C(C)=O
InChI
1S/C6H12O3/c1-5(7)6(2,8-3)9-4/h1-4H3
InChI key
UFQBSPGKRRSATO-UHFFFAOYSA-N
Related Categories
General description
3,3-Dimethoxy-2-butanone is a biacetyl monoketal.
Application
3,3-Dimethoxy-2-butanone may be used as starting reagent in the synthesis of 6-acety1-l,2-dihydro-2-oxo-3-pyridinecarboxylic acid. It may be used in the preparation of 4,4-dimethoxy-3-oxo-1-phenyl-1-pentene.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
113.0 °F - closed cup
Flash Point(C)
45 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Magnesium Methoxide Cyclization of Biacetyl Derivatives1.
Muxfeldt H, et al.
The Journal of Organic Chemistry, 30(10), 3573-3574 (1965)
An efficient synthesis and mechanism of formation of 6-acetyl-1, 2-dihydro-2-oxo-3-pyridinecarboxylic acid.
Sanchez JP.
Tetrahedron, 46(23), 7693-7698 (1990)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service