Skip to Content
MilliporeSigma
All Photos(1)

Documents

341045

Sigma-Aldrich

2-Ethyl-2-methyl-1,3-dioxolane

99%

Synonym(s):

2-Butanone ethylene ketal, 2-Ethyl-2-methyl-1,3-dioxolane, 2-Ethyl-2-methyldioxolane, 2-Methyl-2-ethyl-1,3-dioxolane, 2-Methyl-2-ethyldioxolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O2
CAS Number:
Molecular Weight:
116.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.409 (lit.)

bp

116-117 °C (lit.)

density

0.929 g/mL at 25 °C (lit.)

SMILES string

CCC1(C)OCCO1

InChI

1S/C6H12O2/c1-3-6(2)7-4-5-8-6/h3-5H2,1-2H3

InChI key

UPZFLZYXYGBAPL-UHFFFAOYSA-N

Application

2-Ethyl-2-methyl-1,3-dioxolane was employed as reagent in the selective ketalization of 1-oxo functions of 8a-methyl 1,6-dioxo 1,2,3,4,6,7,8,8a-octahydronaphthalene and 7a-methyl 1,5-dioxo 5,6,7,7a-tetrahydroindane. It was also employed in the enantioselective total synthesis of (-)-strychnine.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Etudes en series bicycliques: Cetalisation selective de dicetones octalinique et tetrahydroindanique et stereochimie de leurs analogues satures.
Bauduin G and Pietrasanta Y.
Tetrahedron, 29(24), 4225-4231 (1973)
Takashi Ohshima et al.
Journal of the American Chemical Society, 124(49), 14546-14547 (2002-12-06)
The enantioselective total synthesis of (-)-strychnine was accomplished through the use of the highly practical catalytic asymmetric Michael reaction (0.1 mol % of (R)-ALB, more than kilogram scale, without chromatography, 91% yield and >99% ee) as well as a tandem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service