Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

339512

Sigma-Aldrich

2-Methylene-1,3-propanediol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH2OH)2
CAS Number:
Molecular Weight:
88.11
Beilstein:
1737653
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.473 (lit.)

bp

93-95 °C/2 mmHg (lit.)

density

1.081 g/mL at 25 °C (lit.)

SMILES string

OCC(=C)CO

InChI

1S/C4H8O2/c1-4(2-5)3-6/h5-6H,1-3H2

InChI key

JFFYKITVXPZLQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hochan Chang et al.
ChemSusChem, 13(19), 5213-5219 (2020-09-24)
Catalytic strategies were developed to synthesize and release chemicals for applications in fine chemicals, such as drugs and polymers, from a biomass-derived chemical, 5-hydroxymethyl furfural (HMF). The combination of the diene and aldehyde functionalities in HMF enabled catalytic production of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service