Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

339393

Sigma-Aldrich

Ethyl 4-methyl-4-pentenoate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CH2CH2CO2C2H5
CAS Number:
Molecular Weight:
142.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

85 °C/20 mmHg (lit.)

density

0.891 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)CCC(C)=C

InChI

1S/C8H14O2/c1-4-10-8(9)6-5-7(2)3/h2,4-6H2,1,3H3

InChI key

RRTBSPBUHUUTHR-UHFFFAOYSA-N

Related Categories

General description

Ethyl 4-methyl-4-pentenoate is an olefin ester. It undergoes iron carbonyl-promoted isomerization to α,β-unsaturated esters. Reaction of ethyl 4-methyl-4-pentenoate with CH3OBOCH3+ was examined in a small Fourier-transform ion cyclotron resonance mass spectrometer equipped with a permanent magnet.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K K Thoen et al.
Journal of the American Society for Mass Spectrometry, 7(11), 1138-1143 (1996-11-01)
The usefulness of CH3OBOCH 3 (+) as a chemical ionization reagent was examined by allowing the ion to react with carboxylic esters of various chain lengths in a small Fourier-transform ion cyclotron resonance mass spectrometer equipped with a permanent magnet.
Kuo-Chen Shih et al.
The Journal of organic chemistry, 61(22), 7784-7792 (1996-11-01)
Ultraviolet photolysis of stoichiometric amounts of methyl oleate and Fe(CO)(5) in hexanes solvent at 0 degrees C gives Fe(CO)(3)(eta(4)-alpha,beta-ester) in which the alpha,beta-unsaturated ester isomer of methyl oleate is stabilized by eta(4)-oxadiene pi coordination of the olefin and ester carbonyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service